3-Fluor-4-(trifluormethyl)benzylbromid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Brommethyl)-3-fluor-4-(trifluormethyl)benzol
PC4307-10G
PC4307-1G
PC4307-5G
PC4307-25G
APOSPC4307-10GEA
106
EUR
APOSPC4307-10G
APOSPC4307-1G
APOSPC4307-5G
APOSPC4307-25G
3-Fluor-4-(trifluormethyl)benzylbromid 97%
3-Fluor-4-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrF₄ Molecular Weight: 257,03 g/mol Schmelzpunkt: 25…27 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236289 CAS-Nummer: 213203-65-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...