Pentafluorophenyl methacrylate
Lieferant: Apollo Scientific
|
Achtung
|
PC4318-1G
PC4318-5G
APOSPC4318-1GEA
49.5
EUR
APOSPC4318-1G
APOSPC4318-5G
Pentafluorophenyl methacrylate
Pentafluorophenyl methacrylate
Formel:
C₁₀H₅F₅O₂ Siedepunkt: 71 °C/2mmHg Dichte: 1,394 g/cm³ (25 °C) Flammpunkt: 77 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042332 CAS-Nummer: 13642-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...