3,4,5-Trifluorphenylboronsäure 97%
Lieferant: Apollo Scientific
Achtung
Synonyme: (3,4,5-Trifluorphenyl)metaborsäure, 3,4,5-Trifluorbenzolboronsäure, Dihydroxy(3,4,5-trifluorphenyl)boran
PC4336-5G PC4336-100G PC4336-25G
APOSPC4336-5GEA 21.9 EUR
APOSPC4336-5G APOSPC4336-100G APOSPC4336-25G
3,4,5-Trifluorphenylboronsäure 97%
3,4,5-Trifluorphenylboronsäure
Very reactive catalyst to produce amides in high yieldsfrom carboxylic acids and primary or secondary aminesJ. Org. Chem., 61, 4196 (1996)
Formel: C₆H₄BF₃O₂
Molecular Weight: 175.9 g/mol
Schmelzpunkt: 275…279 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093069
CAS-Nummer: 143418-49-9