2-Fluor-4-(trifluormethyl)benzamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4371S-1G
PC4371S-25G
PC4371S-5G
APOSPC4371S-1GEA
21.9
EUR
APOSPC4371S-1G
APOSPC4371S-25G
APOSPC4371S-5G
2-Fluor-4-(trifluormethyl)benzamid 97%
2-Fluor-4-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Molecular Weight: 207.13 g/mol Schmelzpunkt: 140…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061147 CAS-Nummer: 207853-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...