2-Fluor-5-(trifluormethyl)benzamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4371T-25G
PC4371T-5G
PC4371T-1G
APOSPC4371T-25GEA
259
EUR
APOSPC4371T-25G
APOSPC4371T-5G
APOSPC4371T-1G
2-Fluor-5-(trifluormethyl)benzamid 98%
2-Fluor-5-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Schmelzpunkt: 115…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061148 CAS-Nummer: 207919-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...