2-Fluor-6-(trifluormethyl)benzamid 97%
Lieferant: Apollo Scientific
PC4371U-5G
PC4371U-10G
PC4371U-1G
APOSPC4371U-5GEA
55.1
EUR
APOSPC4371U-5G
APOSPC4371U-10G
APOSPC4371U-1G
2-Fluor-6-(trifluormethyl)benzamid 97%
2-Fluor-6-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Molecular Weight: 207.13 g/mol Schmelzpunkt: 144…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083555 CAS-Nummer: 144851-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...