3-Fluor-5-(trifluormethyl)benzamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4371V-1G
PC4371V-25G
PC4371V-5G
APOSPC4371V-1GEA
21.9
EUR
APOSPC4371V-1G
APOSPC4371V-25G
APOSPC4371V-5G
3-Fluor-5-(trifluormethyl)benzamid 97%
3-Fluor-5-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Molecular Weight: 207.13 g/mol Schmelzpunkt: 115…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061149 CAS-Nummer: 207986-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...