2-Fluor-3-(trifluormethyl)benzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4372K-25G
PC4372K-5G
APOSPC4372K-25GEA
66.7
EUR
APOSPC4372K-25G
APOSPC4372K-5G
2-Fluor-3-(trifluormethyl)benzaldehyd 97%
2-Fluoro-3-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄F₄O Molecular Weight: 192,11 g/mol Dichte: 1,393 g/cm³ (25 °C) Flammpunkt: 73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061246 CAS-Nummer: 112641-20-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...