α,α,α,6-Tetrafluor-m-toluylsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-(trifluoromethyl)benzoic acid

PC4373A-1G PC4373A-5G PC4373A-25G PC4373A-100G
APOSPC4373A-1GEA 14.7 EUR
APOSPC4373A-1G APOSPC4373A-5G APOSPC4373A-25G APOSPC4373A-100G
α,α,α,6-Tetrafluor-m-toluylsäure 98%
α,α,α,6-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
Schmelzpunkt: 100…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061292
CAS-Nummer: 115029-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR