α,α,α,5-Tetrafluor-m-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluor-5-(trifluormethyl)benzoesäure
PC4373C-100G
PC4373C-25G
PC4373C-5G
APOSPC4373C-100GEA
169
EUR
APOSPC4373C-100G
APOSPC4373C-25G
APOSPC4373C-5G
α,α,α,5-Tetrafluor-m-toluylsäure 98%
α,α,α,5-Tetrafluor-m-toluylsäure
Formel:
C₈H₄F₄O₂ Molecular Weight: 208.11 g/mol Schmelzpunkt: 104…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061293 CAS-Nummer: 161622-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...