α,α,α,4-Tetrafluor-m-toluylsäure 98%

Lieferant: Apollo Scientific
Achtung

PC4373DB-100G PC4373DB-25G PC4373DB-5G
APOSPC4373DB-100GEA 87 EUR
APOSPC4373DB-100G APOSPC4373DB-25G APOSPC4373DB-5G
α,α,α,4-Tetrafluor-m-toluylsäure 98%
α,α,α,4-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061294
CAS-Nummer: 67515-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR