4-Fluor-3-(trifluormethyl)benzophenon 97%

Lieferant: Apollo Scientific
Achtung

PC4374L-25G PC4374L-5G PC4374L-1G
APOSPC4374L-25GEA 299 EUR
APOSPC4374L-25G APOSPC4374L-5G APOSPC4374L-1G
4-Fluor-3-(trifluormethyl)benzophenon 97%
4-Fluor-3-(trifluormethyl)benzophenon
Formel: C₁₄H₈F₄O
Molecular Weight: 268.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061258
CAS-Nummer: 239087-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR