2-Fluor-6-(trifluormethyl)benzylamin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Aminomethyl)-3-fluorobenzotrifluoride, [2-Fluoro-6-(trifluoromethyl)phenyl]methylamine
PC4374W-1G
PC4374W-5G
PC4374W-25G
APOSPC4374W-1GEA
21.9
EUR
APOSPC4374W-1G
APOSPC4374W-5G
APOSPC4374W-25G
2-Fluor-6-(trifluormethyl)benzylamin 95%
2-Fluor-6-(trifluormethyl)benzylamin
Formel:
C₈H₇F₄N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061165 CAS-Nummer: 239087-06-0 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...