2-Fluor-5-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-(trifluoromethyl)phenyl)methanol

PC4375G-25G PC4375G-5G PC4375G-100G
APOSPC4375G-25GEA 69.6 EUR
APOSPC4375G-25G APOSPC4375G-5G APOSPC4375G-100G
2-Fluor-5-(trifluormethyl)benzylalkohol 98%
2-Fluor-5-(trifluormethyl)benzylalkohol
Formel: C₈H₆F₄O
Molecular Weight: 194,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061167
CAS-Nummer: 207974-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR