(2-Fluor-4-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-(trifluoromethyl)phenylacetic acid

PC4376T-5G PC4376T-1G
APOSPC4376T-5GEA 440 EUR
APOSPC4376T-5G APOSPC4376T-1G
(2-Fluor-4-(trifluormethyl)phenyl)essigsäure 98%
(2-Fluor-4-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆F₄O₂
Molecular Weight: 222,14 g/mol
Schmelzpunkt: 78…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061186
CAS-Nummer: 209991-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR