(2-Fluor-5-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid

PC4376U-1G PC4376U-250MG PC4376U-5G PC4376U-25G
APOSPC4376U-1GEA 21.9 EUR
APOSPC4376U-1G APOSPC4376U-250MG APOSPC4376U-5G APOSPC4376U-25G
(2-Fluor-5-(trifluormethyl)phenyl)essigsäure 98%
(2-Fluor-5-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆F₄O₂
Molecular Weight: 222,14 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061187
CAS-Nummer: 220227-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR