(3-Fluor-5-(trifluormethyl)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-5-(trifluoromethyl)phenylacetic Acid
PC4376W-5G
PC4376W-1G
PC4376W-25G
APOSPC4376W-5GEA
55.1
EUR
APOSPC4376W-5G
APOSPC4376W-1G
APOSPC4376W-25G
(3-Fluor-5-(trifluormethyl)phenyl)essigsäure 98%
(3-Fluor-5-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆F₄O₂ Molecular Weight: 222,14 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061188 CAS-Nummer: 195447-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...