3-Fluor-L-tyrosin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-hydroxy-L-phenylalanine, (S)-2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid, H-Tyr(3-F)-OH

PC4378-1G PC4378-250MG
APOSPC4378-1GEA 294 EUR
APOSPC4378-1G APOSPC4378-250MG
3-Fluor-L-tyrosin 97%
3-Fluor-L-tyrosin
Formel: C₉H₁₀FNO₃
Molecular Weight: 199,18 g/mol
Schmelzpunkt: 260…261 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002607
CAS-Nummer: 7423-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR