5-Fluor-m-xylol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dimethyl-5-fluorbenzol, 1-Fluor-3,5-dimethylbenzol, 5-Fluor-1,3-dimethylbenzol
PC4381J-100G
PC4381J-25G
PC4381J-5G
APOSPC4381J-100GEA
65.4
EUR
APOSPC4381J-100G
APOSPC4381J-25G
APOSPC4381J-5G
5-Fluor-m-xylol 98%
5-Fluor-m-xylol
Formel:
C₈H₉F Molecular Weight: 124.16 g/mol Siedepunkt: 144…145 °C Flammpunkt: 35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052366 CAS-Nummer: 461-97-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...