Methyl-3-(trifluoromethyl)phenylacetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-(3-(trifluoromethyl)phenyl)acetate
PC4391-1G
PC4391-25G
PC4391-5G
APOSPC4391-1GEA
21.9
EUR
APOSPC4391-1G
APOSPC4391-25G
APOSPC4391-5G
Methyl-3-(trifluoromethyl)phenylacetate
Methyl-3-(trifluoromethyl)phenylacetate
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218.175 g/mol Siedepunkt: 211 °C Dichte: 1.24 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039762 CAS-Nummer: 62451-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...