Yttrium(III)trifluormethansulfonat

Lieferant: Apollo Scientific

Synonyme: Yttrium(III) trifluoromethanesulphonate, Yttrium triflate

PC4392-10G PC4392-50G PC4392-25G PC4392-100G
APOSPC4392-10GEA 0 EUR
APOSPC4392-10G APOSPC4392-50G APOSPC4392-25G APOSPC4392-100G
Yttrium(III)trifluormethansulfonat
Yttrium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Y
Lagertemperatur: Raumtemperatur
MDL: MFCD00209623
CAS-Nummer: 52093-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR