1,1,1,2,2,3,3-Heptafluor-4-iodbutan 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H,1H-Heptafluorobutyl iodide, 2,2,3,3,4,4,4-Heptafluoro-1-iodobutane
PC4400-1G
PC4400-5G
APOSPC4400-1GEA
94.4
EUR
APOSPC4400-1G
APOSPC4400-5G
1,1,1,2,2,3,3-Heptafluor-4-iodbutan 97%
1,1,1,2,2,3,3-Heptafluor-4-iodbutan
Formel:
C₄H₂F₇I Molecular Weight: 309,95 g/mol Siedepunkt: 91…92 °C Dichte: 2,054 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00013719 CAS-Nummer: 374-98-1 EINECS: 206-779-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...