4-Amino-3-fluorbenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Amino-3-fluorophenyl)methanol
PC4408-1G
PC4408-250MG
PC4408-5G
APOSPC4408-1GEA
128
EUR
APOSPC4408-1G
APOSPC4408-250MG
APOSPC4408-5G
4-Amino-3-fluorbenzylalkohol
4-Amino-3-fluorbenzylalkohol
Formel:
C₇H₈FNO Molecular Weight: 141.1429 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03094241 CAS-Nummer: 146019-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...