1H,1H-Perfluorohexan-1-ol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H,1H-Undecafluoro-1-hexanol, 1H,1H-Perfluoro-1-hexanol, 1,1-Dihydroperfluorohexanol

PC4438-25G PC4438-5G
APOSPC4438-25GEA 719 EUR
APOSPC4438-25G APOSPC4438-5G
1H,1H-Perfluorohexan-1-ol 98%
1H,1H-Perfluorohexan-1-ol
Formel: C₆H₃F₁₁O
Molecular Weight: 300,07 g/mol
Siedepunkt: 130…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038338
CAS-Nummer: 423-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR