4-Brom-2-fluor-3-methylpyridin 98%

Lieferant: Apollo Scientific
Achtung

PC445018-1G PC445018-5G PC445018-25G
APOSPC445018-1GEA 104 EUR
APOSPC445018-1G APOSPC445018-5G APOSPC445018-25G
4-Brom-2-fluor-3-methylpyridin 98%
4-Bromo-2-fluoro-3-methylpyridine
Formel: C₆H₅BrFN
Molecular Weight: 190,0130 g/mol
Siedepunkt: 60 °C/3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD09839278
CAS-Nummer: 128071-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR