2-Chlor-3-fluor-4-iodpyridin 95+%

Lieferant: Apollo Scientific

PC445020-25G
APOSPC445020-25GEA 142 EUR
APOSPC445020-25G
2-Chlor-3-fluor-4-iodpyridin 95+%
2-Chloro-3-fluoro-4-iodopyridine
Formel: C₅H₂ClFIN
Molecular Weight: 257,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972389
CAS-Nummer: 148639-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR