4-Fluor-3-methylpyridin 97%
Lieferant: Apollo Scientific
PC445030-250MG
PC445030-1G
PC445030-5G
APOSPC445030-250MGEA
107
EUR
APOSPC445030-250MG
APOSPC445030-1G
APOSPC445030-5G
4-Fluor-3-methylpyridin 97%
4-Fluor-3-methylpyridin
Formel:
C₆H₆FN Molecular Weight: 111.12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04114241 CAS-Nummer: 28489-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...