3-Brom-2-fluor-6-methylpyridin
Lieferant: Apollo Scientific
Synonyme:
3-Brom-2-fluor-6-picolin
PC445031-1G
PC445031-25G
PC445031-5G
APOSPC445031-1GEA
94.5
EUR
APOSPC445031-1G
APOSPC445031-25G
APOSPC445031-5G
3-Brom-2-fluor-6-methylpyridin
3-Bromo-2-fluoro-6-methylpyridine
Formel:
C₆H₅BrFN Molecular Weight: 190.01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095080 CAS-Nummer: 375368-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...