2-Chlor-5-fluor-3-hydroxypyridin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-fluoropyridin-3-ol

PC445038-100MG PC445038-250MG
APOSPC445038-100MGEA 60.9 EUR
APOSPC445038-100MG APOSPC445038-250MG
2-Chlor-5-fluor-3-hydroxypyridin 98%
2-Chloro-5-fluoro-3-hydroxypyridine
Formel: C₆H₄ClFO
Molecular Weight: 147,5400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095268
CAS-Nummer: 884494-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR