3-Brom-2-chlor-5-fluorpyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC445039-25G
APOSPC445039-25GEA
108
EUR
APOSPC445039-25G
3-Brom-2-chlor-5-fluorpyridin 95%
3-Bromo-2-chloro-5-fluoropyridine
Formel:
C₅H₂BrClFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095269 CAS-Nummer: 884494-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...