2-Chlor-5-fluor-4-iodpyridin 98%

Lieferant: Apollo Scientific
Achtung

PC445042-5G
APOSPC445042-5GEA 108 EUR
APOSPC445042-5G
2-Chlor-5-fluor-4-iodpyridin 98%
2-Chloro-5-fluoro-4-iodopyridine
Formel: C₅H₂ClFIN
Molecular Weight: 257,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095306
CAS-Nummer: 884494-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR