4-Fluor-2-methoxybenzylamin 95+%

Lieferant: Apollo Scientific

Synonyme: (4-Fluoro-2-methoxyphenyl)methanamine, (4-Fluoro- 2-methoxy-phenyl) methanamine

PC445061-1G
APOSPC445061-1GEA 120 EUR
APOSPC445061-1G
4-Fluor-2-methoxybenzylamin 95+%
4-Fluor-2-methoxybenzylamin
Formel: C₈H₁₀FNO
Molecular Weight: 155.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116361
CAS-Nummer: 870563-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR