N'-[1-Amino-2-(3-(trifluoromethyl)phenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Lieferant: Apollo Scientific
PC446002-1G
PC446002-500MG
APOSPC446002-1GEA
183
EUR
APOSPC446002-1G
APOSPC446002-500MG
N'-[1-Amino-2-(3-(trifluoromethyl)phenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(3-(trifluoromethyl)phenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formel:
C₁₄H₁₈F₃N₃O₃ Molecular Weight: 333,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10568188 CAS-Nummer: 1053655-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...