N'-[1-Amino-2-(3,5-bis-(trifluoromethyl)phenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Lieferant: Apollo Scientific

PC446006-1G PC446006-500MG
APOSPC446006-1GEA 183 EUR
APOSPC446006-1G APOSPC446006-500MG
N'-[1-Amino-2-(3,5-bis-(trifluoromethyl)phenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(3,5-bis-(trifluoromethyl)phenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formel: C₁₅H₁₇F₆N₃O₂
Molecular Weight: 385,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568198
CAS-Nummer: 1053656-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR