Ethyl-5-(3-(trifluoromethyl)phenyl)[1,2,4]oxadiazole-3-carboxylate

Lieferant: Apollo Scientific

PC446008-1G PC446008-5G PC446008-10G
APOSPC446008-1GEA 168 EUR
APOSPC446008-1G APOSPC446008-5G APOSPC446008-10G
Ethyl-5-(3-(trifluoromethyl)phenyl)[1,2,4]oxadiazole-3-carboxylate
Ethyl-5-(3-(trifluoromethyl)phenyl)[1,2,4]oxadiazole-3-carboxylate
Formel: C₁₂H₉F₃N₂O₃
Molecular Weight: 286.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568215
CAS-Nummer: 163720-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR