2-(Trifluoromethoxy)benzamidine hydrochloride

Lieferant: Apollo Scientific

PC446018-1G PC446018-5G PC446018-10G
APOSPC446018-1GEA 154 EUR
APOSPC446018-1G APOSPC446018-5G APOSPC446018-10G
2-(Trifluoromethoxy)benzamidine hydrochloride
2-(Trifluoromethoxy)benzamidine hydrochloride
Formel: C₈H₈ClF₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00173664
CAS-Nummer: 127979-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR