4-Methyl-6-(trifluoromethyl)pyrimidin-2-amine 95+%
Lieferant: Apollo Scientific
PC446095-1G
APOSPC446095-1GEA
311
EUR
APOSPC446095-1G
4-Methyl-6-(trifluoromethyl)pyrimidin-2-amine 95+%
4-Methyl-6-(trifluoromethyl)pyrimidin-2-amine
Formel:
C₆H₆F₃N₃ Molecular Weight: 177,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160470 CAS-Nummer: 5734-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...