2-Brom-3',5'-difluoracetophenon 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(3,5-difluorophenyl)ethanone, 3,5-Difluorophenacyl bromide

PC4473-1G PC4473-5G
APOSPC4473-1GEA 102 EUR
APOSPC4473-1G APOSPC4473-5G
2-Brom-3',5'-difluoracetophenon 98%
2-Brom-3',5'-difluoracetophenon
Formel: C₈H₅BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03094245
CAS-Nummer: 220607-75-0
UN: 2920
ADR: 8+3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR