4-(Difluormethoxy)benzolsulfonamid

Lieferant: Apollo Scientific

PC448011-1G PC448011-5G PC448011-25G
APOSPC448011-1GEA 66.7 EUR
APOSPC448011-1G APOSPC448011-5G APOSPC448011-25G
4-(Difluormethoxy)benzolsulfonamid
4-(Difluormethoxy)benzolsulfonamid
Formel: C₇H₇F₂NO₃S
Molecular Weight: 223,2000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407973
CAS-Nummer: 874781-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR