5-Fluoro-1H-indazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

PC448014-1G PC448014-5G
APOSPC448014-1GEA 161 EUR
APOSPC448014-1G APOSPC448014-5G
5-Fluoro-1H-indazole-3-carboxylic acid 95%
5-Fluoro-1H-indazole-3-carboxylic acid
Formel: C8H5FN2O2
Molecular Weight: 180,140 g/mol
Schmelzpunkt: 299 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03840628
CAS-Nummer: 1077-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR