Trifluoromethanesulphonanilide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N-Phenyl(trifluoromethane)sulphonamide, 1,1,1-Trifluoro-N-phenylmethanesulphonamide
PC448034-1G
PC448034-5G
PC448034-25G
APOSPC448034-1GEA
21.9
EUR
APOSPC448034-1G
APOSPC448034-5G
APOSPC448034-25G
Trifluoromethanesulphonanilide
Trifluoromethanesulphonanilide
Formel:
C₇H₆F₃NO₂S Molecular Weight: 225.19 g/mol Schmelzpunkt: 67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00521820 CAS-Nummer: 456-64-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...