4-Brom-2-chlor-6-fluorphenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4481-1G
PC4481-5G
PC4481-25G
PC4481-100G
APOSPC4481-1GEA
21.9
EUR
APOSPC4481-1G
APOSPC4481-5G
APOSPC4481-25G
APOSPC4481-100G
4-Brom-2-chlor-6-fluorphenol 97%
4-Brom-2-chlor-6-fluorphenol
Formel:
C₆H₃BrClFO Molecular Weight: 225,44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631476 CAS-Nummer: 161045-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...