1,1,1,2,2,3,3-Heptafluoro-5-iodopentane 94%
Lieferant: Apollo Scientific
|
Achtung
|
PC4496U-5G
PC4496U-1G
PC4496U-25G
APOSPC4496U-5GEA
92.8
EUR
APOSPC4496U-5G
APOSPC4496U-1G
APOSPC4496U-25G
1,1,1,2,2,3,3-Heptafluoro-5-iodopentane 94%
1,1,1,2,2,3,3-Heptafluoro-5-iodopentane
Formel:
C₅H₄F₇I Molecular Weight: 323,98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155834 CAS-Nummer: 1513-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...