(1,1-Difluoropropyl)benzene

Lieferant: Apollo Scientific
Achtung

PC450049-1G PC450049-5G
APOSPC450049-1GEA 205 EUR
APOSPC450049-1G APOSPC450049-5G
(1,1-Difluoropropyl)benzene
(1,1-Difluoropropyl)benzene
Formel: C₉H₁₀F₂
Molecular Weight: 156.18 g/mol
Siedepunkt: 25 °C/2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD14525542
CAS-Nummer: 74185-83-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR