Phenyl trifluoromethanesulphonate 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Phenyl Trifluoromethanesulphonate, Trifluoromethanesulphonic acid Phenyl Ester, Phenyl Triflate

PC450234-25G PC450234-5G
APOSPC450234-25GEA 263 EUR
APOSPC450234-25G APOSPC450234-5G
Phenyl trifluoromethanesulphonate 98%
Phenyl trifluoromethanesulphonate
Formel: C₇H₅F₃O₃S
Molecular Weight: 226,17 g/mol
Siedepunkt: 99…100 °C
Dichte: 1,396 g/cm³ (25 °C)
Flammpunkt: 71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192399
CAS-Nummer: 17763-67-6
UN: 2927
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR