Pyridinium trifluoromethanesulphonate

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyridinium Triflate

PC450250-100G PC450250-25G PC450250-5G
APOSPC450250-100GEA 259 EUR
APOSPC450250-100G APOSPC450250-25G APOSPC450250-5G
Pyridinium trifluoromethanesulphonate
Pyridinium trifluoromethanesulphonate
Formel: C₆H₆F₃NO₃S
Molecular Weight: 229,17 g/mol
Schmelzpunkt: 221…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00274656
CAS-Nummer: 52193-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR