Enoxacin

Lieferant: Apollo Scientific

Synonyme: 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic acid

PC450274-25G PC450274-5G
APOSPC450274-25GEA 328 EUR
APOSPC450274-25G APOSPC450274-5G
Enoxacin
Enoxacin
Formel: C₁₅H₁₇FN₄O₃
Molecular Weight: 320,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00133308
CAS-Nummer: 74011-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR