(2-Trifluoromethylphenyl)methanesulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Trifluoromethyl)phenyl)methanesulphonamide
PC450275-100G
PC450275-25G
PC450275-5G
APOSPC450275-100GEA
1080
EUR
APOSPC450275-100G
APOSPC450275-25G
APOSPC450275-5G
(2-Trifluoromethylphenyl)methanesulphonamide
(2-Trifluoromethylphenyl)methanesulphonamide
Formel:
C₈H₈F₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09763601 CAS-Nummer: 112941-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...