1,1,1,2,2,3,3-Heptafluor-3-[(trifluorvinyl)oxy]propan 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (Heptafluorpropoxy)trifluorethen, (1,1,2,2,3,3,3-Heptafluorpropoxy)trifluorethen

PC4513L-100G PC4513L-25G
APOSPC4513L-100GEA 498 EUR
APOSPC4513L-100G APOSPC4513L-25G
1,1,1,2,2,3,3-Heptafluor-3-[(trifluorvinyl)oxy]propan 97%
1,1,1,2,2,3,3-Heptafluor-3-[(trifluorvinyl)oxy]propan

Useful intermediate and monomer; the co-polymer with tetrafluoroethylene is sold as Teflon PFA.

Formel: C₅F₁₀O
Molecular Weight: 266,04 g/mol
Siedepunkt: 35…36 °C
Schmelzpunkt: –70 °C
Dichte: 1,53 g/cm³ (25 °C)
Flammpunkt: –20 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00191577
CAS-Nummer: 1623-05-8
EINECS: 216-600-2
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR