3-Fluorphenethylbromid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Bromoethyl)-3-fluorobenzene
PC4528-25G
PC4528-5G
PC4528-250MG
APOSPC4528-25GEA
219
EUR
APOSPC4528-25G
APOSPC4528-5G
APOSPC4528-250MG
3-Fluorphenethylbromid 95%
3-Fluorphenethylbromid
Formel:
C₈H₈BrF Molecular Weight: 203,05 g/mol Siedepunkt: 94 °C/10mmHg Dichte: 1,45 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095397 CAS-Nummer: 25017-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...